missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid Tris(2-methylpropyl) Ester, TRC

Product Code. 30426311
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426311 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426311 Supplier Toronto Research Chemicals Supplier No. TRCP35947510G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphoric Acid Tris(2-methylpropyl) Ester
CAS 126-71-6
Molecular Formula C12 H27 O4 P
Quantity 10 g
Synonym Phosphoric acid, tris(2-methylpropyl) ester, Isobutyl phosphate ((C4H9O)3PO) (6CI,7CI), Phosphoric acid, triisobutyl ester (8CI), Antifoam TIP, Daiguard 400, NSC 62222, Reomol TIBP, Triisobutyl phosphate
InChI Formula InChI=1S/C12H27O4P/c1-10(2)7-14-17(13,15-8-11(3)4)16-9-12(5)6/h10-12H,7-9H2,1-6H3
SMILES CC(C)COP(=O)(OCC(C)C)OCC(C)C
IUPAC Name tris(2-methylpropyl) phosphate
Molecular Weight (g/mol) 266.31
Formula Weight 266.1647
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.