missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid Tris(2-methylpropyl) Ester, TRC

Product Code. 30008047
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008047 1 g 1g
30009704 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008047 Supplier Toronto Research Chemicals Supplier No. TRCP3594751G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-71-6
Molecular Formula C12 H27 O4 P
Molecular Weight (g/mol) 266.31
Synonym Phosphoric acid, tris(2-methylpropyl) ester, Isobutyl phosphate ((C4H9O)3PO) (6CI,7CI), Phosphoric acid, triisobutyl ester (8CI), Antifoam TIP, Daiguard 400, NSC 62222, Reomol TIBP, Triisobutyl phosphate
IUPAC Name tris(2-methylpropyl) phosphate
SMILES CC(C)COP(=O)(OCC(C)C)OCC(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H27O4P/c1-10(2)7-14-17(13,15-8-11(3)4)16-9-12(5)6/h10-12H,7-9H2,1-6H3
Formula Weight 266.1647
Chemical Name or Material Phosphoric Acid Tris(2-methylpropyl) Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.