missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid Silver Salt, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Phosphoric Acid Silver Salt |
| CAS | 7784-09-0 |
| Molecular Formula | Ag3O4P |
| Quantity | 10 g |
| Synonym | Silver Phosphate, Silver Orthophosphate, Silver Orthophosphate (Ag3PO4), Silver Phosphate (Ag3PO4), Trisilver Phosphate, Trisilver(1+) Orthophosphate, Trisilver(1+) Phosphate |
| InChI Formula | InChI=1S/3Ag.H3O4P/c;;;1-5(2,3)4/h;;;(H3,1,2,3,4)/q3*+1;/p-3 |
| SMILES | [Ag+].[Ag+].[Ag+].[O-]P(=O)([O-])[O-] |
| IUPAC Name | trisilver;phosphate |
| Molecular Weight (g/mol) | 418.58 |
| Formula Weight | 415.6687 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction