missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric acid, Puriss. p.a., crystallized, ≥99.0% (T), Solstice

Product Code. 15607160 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass Bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15607160 100 g Glass Bottle 100g
15627160 500 g Glass Bottle 500g
15617160 1 kg Glass Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15607160 Supplier Honeywell-Fluka Supplier No. 79622100G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., crystallized,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Melting Point ∼40°C (literature)
Boiling Point 158°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Density 1.685 g/mL at 25 °C(lit.)
Packaging Glass Bottle
Vapor Pressure 2.2 mmHg (20°C), 5 mmHg (25°C)
Quantity 100 g
Linear Formula H3PO4
UN Number UN3453
Beilstein 1921286
Formula Weight 98
Percent Purity ≥99%
Grade puriss. p.a.
Chemical Name or Material Phosphoric acid, Puriss. p.a., crystallized, ≥99.0% (T)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.