missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric acid, Puriss. p.a., crystallized, ≥99.0% (T), Solstice

Product Code. 16645923
Change view
Click to view available options
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16645923 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16645923 Supplier Fluka Supplier No. 7962225KG
  • ONLINE EXCLUSIVE PRICE
    £2275.00
    £2096.10 / 25kg
    Save £178.90
    7% Off
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Colorless
CAS 7664-38-2
Melting Point 42°C
CAS Max % ≤ 100%
Density 1.880 g/cm3 (20°C)
Quantity 25 kg
Molecular Formula H3O4P
Linear Formula H3PO4
Beilstein 1921286
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILES OP(O)(O)=O
IUPAC Name phosphoric acid
Molecular Weight (g/mol) 97.99
Grade Reagent
Chemical Name or Material Phosphoric acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.