missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric acid, Puriss., meets analytical spec. of BP, FCC, NF, Ph. Eur., 85.0-88.0%, Solstice

Product Code. 15680710 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
4 x 2.5 L
45 kg
4 x 5 L
6 x 1 L
Unit Size:
1 L
10 L
2.5 L
45kg
5 L
Pack of 4
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15680710 4 x 5 L Pack of 4
15640710 1 L 1 L
15650710 2.5 L 2.5 L
15690710 5 L 5 L
15600720 6 x 1 L Pack of 6
15670710 4 x 2.5 L Pack of 4
15630710 10 L 10 L
15660710 45 kg 45kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15680710 Supplier Honeywell-Fluka Supplier No. 041074X5L
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, FCC, NF, Ph. Eur., 85.0-88.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Density 1.685g/mL at 25°C
Vapor Pressure 5mmHg (25°C), 2.2mmHg (20°C)
Quantity 4 x 5 L
Linear Formula H3PO4
UN Number UN1805
Formula Weight 98
Percent Purity 85.0 to 88.0%
Grade puriss.
Chemical Name or Material Phosphoric Acid, Meets analytical specification of Ph. Eur.BP, NF, FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.