missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric acid, Puriss., meets analytical spec. of BP, FCC, NF, Ph. Eur., 85.0-88.0%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
4 x 2.5 L
45 kg
4 x 5 L
6 x 1 L
Unit Size:
1 L
10 L
2.5 L
45kg
5 L
Pack of 4
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Density | 1.685g/mL at 25°C |
| Vapor Pressure | 5mmHg (25°C), 2.2mmHg (20°C) |
| Quantity | 6 x 1 L |
| Linear Formula | H3PO4 |
| UN Number | UN1805 |
| Formula Weight | 98 |
| Percent Purity | 85.0 to 88.0% |
| Grade | puriss. |
| Chemical Name or Material | Phosphoric Acid, Meets analytical specification of Ph. Eur.BP, NF, FCC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction