missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid NF/FCC, Macron Fine Chemicals™
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass
Glass Bottle
Specifications
Packaging | Glass |
Quantity | 500 mL |
Formula Weight | 98 |
Grade | FCC/NF |
Chemical Name or Material | Phosphoric Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'