missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid Monosodium Salt Dihydrate, TRC

Product Code. 30010322
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010322 25 g 25g
30006423 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010322 Supplier Toronto Research Chemicals Supplier No. TRCP35922525G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-35-0
Molecular Formula H2NaO4P .2H2O
Molecular Weight (g/mol) 156.01
Synonym Phosphoric Acid Sodium Salt Hydrate (1:1:2), Monosodium Dihydrogen Phosphate Dihydrate, Monosodium Phosphate Dihydrate, Sodium Dihydrogen Phosphate Dihydrate, Sodium Phosphate Monobasic Dihydrate
IUPAC Name sodium;dihydrogen phosphate;dihydrate
SMILES O.O.[Na+].OP(=O)(O)[O-]

Specifications

Quantity 25 g
InChI Formula InChI=1S/Na.H3O4P.2H2O/c;1-5(2,3)4;;/h;(H3,1,2,3,4);2*1H2/q+1;;;/p-1
Formula Weight 155.98
Chemical Name or Material Phosphoric Acid Monosodium Salt Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.