missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid Dipotassium, TRC

Product Code. 30133368
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133368 100 g 100g
30010308 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133368 Supplier Toronto Research Chemicals Supplier No. TRCP359215100G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-11-4
Synonym Dibasic Potassium Phosphate (AS), Phosphoric Acid Dipotassium Salt, Conclyte P, DKP, Dibasic Potassium Phosphate, Dipotassium Hydrogen Orthophosphate, Dipotassium Hydrogen Phosphate, Dipotassium Hydrogen Phosphate (K2HPO4), Dipotassium Monohydrogen Phosphate, Dipotassium Monophosphate, Dipotassium Orthophosphate, Dipotassium Phosphate, Dipotassium Phosphate (K2HPO4), Hydrogen Dipotassium Phosphate, Potassium Biphosphate, Potassium Dibasic Phosphate (K2HPO4), Potassium Hydrogen Phosphate (K2HPO4), Potassium Monoacid Phosphate, Potassium Monohydrogen Phosphate, Potassium Monophosphate, Potassium Phosphate (K2HPO4), Potassium Phosphate Dibasic, Rhodiaphos DKP, Phosphoric Acid Dipotassium
IUPAC Name dipotassium;hydrogen phosphate
SMILES [K+].[K+].OP(=O)([O-])[O-]

Specifications

Quantity 100 g
InChI Formula InChI=1S/2K.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2
Formula Weight 173.8887
Chemical Name or Material Dipotassium phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.