missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid Diethyl Ester, TRC

Product Code. 30137506
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137506 500 mg 500mg
30133175 1 g 1g
17914635 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137506 Supplier Toronto Research Chemicals Supplier No. TRCP359430500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-02-7
Molecular Formula C4 H11 O4 P
Molecular Weight (g/mol) 154.1
Synonym Phosphoric acid, diethyl ester, Phosphoric acid diethyl ester, Ethyl phosphate ((C2H5O)2(HO)PO) (6CI), DEP, Diethyl acid phosphate, Diethyl hydrogen phosphate, Diethyl phosphate, Diethyl phosphoric acid, JP 502, NSC 97178, O,O-Diethyl hydrogen phosphate
IUPAC Name diethyl hydrogen phosphate
SMILES CCOP(=O)(O)OCC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C4H11O4P/c1-3-7-9(5,6)8-4-2/h3-4H2,1-2H3,(H,5,6)
Formula Weight 154.0395
Percent Purity >95%
Chemical Name or Material Diethyl hydrogen phosphate (as free acid)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.