missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid Dibenzyl Ester, TRC

Product Code. 30007645
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007645 10 g 10g
30006617 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007645 Supplier Toronto Research Chemicals Supplier No. TRCP35940010G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1623-08-1
Molecular Formula C14H15O4P
Molecular Weight (g/mol) 278.24
Synonym Dibenzyl Hydrogen Phosphate, Dibenzyl Phosphate, Dibenzylphosphoric Acid
IUPAC Name dibenzyl hydrogen phosphate
SMILES OP(=O)(OCc1ccccc1)OCc2ccccc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H15O4P/c15-19(16,17-11-13-7-3-1-4-8-13)18-12-14-9-5-2-6-10-14/h1-10H,11-12H2,(H,15,16)
Formula Weight 278.0708
Chemical Name or Material Phosphoric Acid Dibenzyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.