missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric acid, 85% aq. soln.

Product Code. 11469216
Change view
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
Unit Size:
10000mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469216 2500 mL 2500mL
11459216 500 mL 500mL
11479216 10,000 mL 10000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469216 Supplier Thermo Scientific Alfa Aesar Supplier No. A18067.0F
  • ONLINE EXCLUSIVE PRICE
    £97.10
    £83.65 / 2500mL
    Save £13.45
    13% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7664-38-2 | H3O4P | 97.994 g/mol

Phosphoric acid is used mainly in fertilizers. It is used as an external standard for phosphorus-31 Nuclear magnetic resonance (NMR); as an electrolyte in phosphoric acid fuel cells and as a catalyst in the hydration of alkenes to produce alcohols, predominantly ethanol. It finds application as a buffer agent in biology and chemistry; as a solution for anodizing and as a cleaner by construction trades to remove mineral deposits, cementitious smears and hard water stains. It is also used as a pH adjuster in cosmetics and skin-care products and a dispersing agent in detergents and leather treatment. It plays an important role in the preparation of albumin derivatives. It is an important starting material used in the preparation of disodium hydrogen phosphate, trisodium phosphate, calcium dihydrogen phosphate and monohydrogen phosphate. It is also used in dentistry and orthodontics as an etching solution as well as a sanitizing agent in dairy, food and brewing industries. It serves as an acidic fruitlike flavoring in food products. It is widely used to remove rust in steel tools or surfaces by converting the rusted ion in to water soluble phosphate compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphoric acid is used mainly in fertilizers. It is used as an external standard for phosphorus-31 Nuclear magnetic resonance (NMR); as an electrolyte in phosphoric acid fuel cells and as a catalyst in the hydration of alkenes to produce alcohols, predominantly ethanol. It finds application as a buffer agent in biology and chemistry; as a solution for anodizing and as a cleaner by construction trades to remove mineral deposits, cementitious smears and hard water stains. It is also used as a pH adjuster in cosmetics and skin-care products and a dispersing agent in detergents and leather treatment. It plays an important role in the preparation of albumin derivatives. It is an important starting material used in the preparation of disodium hydrogen phosphate, trisodium phosphate, calcium dihydrogen phosphate and monohydrogen phosphate. It is also used in dentistry and orthodontics as an etching solution as well as a sanitizing agent in dairy, food and brewing industries. It serves as an acidic fruitlike flavoring in food products. It is widely used to remove rust in steel tools or surfaces by converting the rusted ion in to water soluble phosphate compound.

Notes
Incompatible with strong bases and powdered metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Melting Point 19°C to 21°C
Concentration 85% aq. soln.
Density 1.7
Linear Formula H3PO4
UN Number UN1805
Quantity 2500 mL
Merck Index 14,7344
Solubility Information Miscible with water,ethanol and mixture of ether: alcohol (3:1).
Formula Weight 98
Chemical Name or Material Phosphoric acid
compliance-icons
Product Identifier
  • Phosphoric acid, 85% aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: orthophosphoric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.