missing translation for 'onlineSavingsMsg'
Learn More

Phosphononitrilic Chloride Trimer, TRC

Product Code. 30419653
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419653 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419653 Supplier Toronto Research Chemicals Supplier No. TRCP35375025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphononitrilic Chloride Trimer
CAS 940-71-6
Molecular Formula Cl6N3P3
Quantity 25 g
Synonym 2,2,4,4,6,6-Hexachloro-2lambda5,4lambda5,6lambda5-1,3,5,2,4,6-triazatriphosphorine, 2,2,4,4,6,6-Hexachloro-2,2,4,4,6,6-hexahydro-1,3,5,2,4,6-triazatriphosphorine, Phosphonitrile Chloride Trimer, 2,2,4,4,6,6-Hexachlorocyclotriphosphazatriene, 2,2,4,4,6,6-Hexachlorocyclotriphosphazene, Cyclophosphazene Dichloride Trimer, Cyclophosphonitrilic Chloride Trimer, Dichlorocyclotriphosphazene, Dichlorophosphazene Cyclic Trimer, Dichlorophosphazene Trimer, Hexachloro-1,3,5,2,4,6-triazatriphosphorine, Hexachlorocyclophosphazatriene, Hexachlorocyclotriphosphatriazene, Hexachlorocyclotriphosphazatriene, Hexachlorocyclotriphosphazene, Hexachlorotriphosphazene, Hexachlorotriphosphonitrile, NSC 209799, NSC 2667, Phosnic 390, Phosphonitrile Chloride Cyclic Trimer, Phosphonitrilic Chloride Cyclic Trimer, Phosphonitrilic Chloride Trimer, Phosphononitrilic Chloride Trimer, Triphosphonitrile Chloride, Triphosphonitrilic Chloride, Tris(phosphonitrilic chloride), Tris(phosphonitriliic chloride)
InChI Formula InChI=1S/Cl6N3P3/c1-10(2)7-11(3,4)9-12(5,6)8-10
SMILES Clp1(Cl)np(Cl)(Cl)np(Cl)(Cl)n1
IUPAC Name 2,2,4,4,6,6-hexachloro-1,3,5-triaza-2λ^{5},4λ^{5},6λ^{5}-triphosphacyclohexa-1,3,5-triene
Molecular Weight (g/mol) 347.66
Formula Weight 344.7436
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.