missing translation for 'onlineSavingsMsg'
Learn More

Phosphonoacetic acid, 98+%

Product Code. 11444030
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444030 10 g 10g
11440053 25 g 25g
11450053 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444030 Supplier Thermo Scientific Alfa Aesar Supplier No. A12117.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4408-78-0 | C2H5O5P | 140.03 g/mol

Phosphonoacetic acid inhibits deoxyribo nucleic acid polymerase, especially in viruses. It is used as an antiviral agent. Further, it is used in the preparation of 1-(dimethylamino)-2,2-bis[(dimethylimmonio)-methyl]ethene diperchlorate by reacting with phosphorus oxychloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphonoacetic acid inhibits deoxyribo nucleic acid polymerase, especially in viruses. It is used as an antiviral agent. Further, it is used in the preparation of 1-(dimethylamino)-2,2-bis[(dimethylimmonio)-methyl]ethene diperchlorate by reacting with phosphorus oxychloride.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4408-78-0
Molecular Formula C2H5O5P
Molecular Weight (g/mol) 140.03
MDL Number MFCD00004311
InChI Key XUYJLQHKOGNDPB-UHFFFAOYSA-N
Synonym phosphonoacetic acid, fosfonet, phosphonoacetate, acetic acid, phosphono, fosfonet sodium, phosphonacetic acid, carboxymethanephosphonic acid, fosfonoacetic acid, disodium phosphonoacetate, lopac-p-6909
PubChem CID 546
ChEBI CHEBI:15732
IUPAC Name 2-phosphonoacetic acid
SMILES OC(=O)CP(O)(O)=O

Specifications

Melting Point 139°C to 146°C
Odor Odorless
Quantity 10 g
UN Number UN3261
Beilstein 1764355
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 140.04
Percent Purity ≥98%
Chemical Name or Material Phosphonoacetic acid
compliance-icons
Product Identifier
  • Phosphonoacetic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Phosphonoacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.