missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphomolybdotungstic Reagent Dilute EU Pharmacopoeia, Fisher Chemical™
Shop All Fisher Chemical Products

Specifications
Specifications
| Packaging | Bottle |
| Analyte or Component Names | Phosphomolybdotungstic Reagent Dilute |
| Purity Grade Notes | EU Ph Ref: 1065001 |
| Element | Phosphomolybdotungstic |
| Quantity | 100 mL |
| Grade | Eu. Ph. |
Safety and Handling
Product Identifier
- Dilute Phosphomolybdotungstic Reagent. European Pharmacopoeia Ref: 1065001 (RP 154)
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?