missing translation for 'onlineSavingsMsg'
Learn More

Phosphomolybdic acid, ammonium salt hydrate

Product Code. 10137763 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10137763 250 g Plastic bottle 250g
10420161 50 g Plastic bottle 50g
10678824 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10137763 Supplier Thermo Scientific Acros Supplier No. 317102500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54723-94-3 | H80Mo12N3O41P-6 | 1960.994 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54723-94-3
Molecular Formula H80Mo12N3O41P-6
Molecular Weight (g/mol) 1960.994
MDL Number MFCD00150423
InChI Key BIQVMZMYTFVCJZ-UHFFFAOYSA-H
Synonym ammonium phosphomolybdate hydrate
PubChem CID 131664298
IUPAC Name azane;molybdenum;trihydroxide;phosphate;tetratriacontahydrate
SMILES N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].[OH-].[OH-].[O-]P(=O)([O-])[O-].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo]

Specifications

Color Yellow
Packaging Plastic bottle
Quantity 250 g
Linear Formula (NH4)3PMo12O40·xH2O
Physical Form Powder
Solubility Information Solubility in water: insoluble.
Formula Weight 1876.35
Chemical Name or Material Phosphomolybdic acid, ammonium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.