missing translation for 'onlineSavingsMsg'
Learn More

Phosphomolybdic Acid (ACS Grade), MP Biomedicals

Product Code. 11447832 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447832 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11447832 Supplier MP Biomedicals Supplier No. 0215255605
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 51429-74-4
Molecular Formula H3Mo12O40P
MDL Number MFCD00149913
Quantity 5 g
InChI Key FEFSFHGZSNHJEL-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].OP(O)(O)=O
IUPAC Name 12-Molybdophosphoric acid
Molecular Weight (g/mol) 1825.40
Formula Weight 3939.5
Grade ACS
Chemical Name or Material Phosphomolybdic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.