missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholipase D2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35105-100ul
This item is not returnable.
View return policy
Description
Phospholipase D2 Polyclonal antibody specifically detects Phospholipase D2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Phospholipase D2 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, ELISA | |
| Choline phosphatase 2, EC 3.1.4.4, hPLD2, Phosphatidylcholine-hydrolyzing phospholipase D2, phospholipase D2, PLD 2, PLD1C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human Phospholipase D2 (NP_002654.3).,, Sequence:, MTATPESLFPTGDELDSSQLQMESDEVDTLKEGEDPADRMHPFLAIYELQSLKVHPLVFAPGVPVTAQVVGTERYTSGSKVGTCTLYSVRLTHGDFSWTTKKKYRHFQELHRDLLRHKVLMSLLPLARFAVAYSPARDAGNREMPSLPRAGPEGSTRHAASKQKYLENYLNRLLTMSFYR | |
| 100 μL | |
| Signal Transduction | |
| 5338 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction