missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholipase C delta 1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Phospholipase C delta 1 Polyclonal antibody specifically detects Phospholipase C delta 1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Classification | Polyclonal |
| Formulation | 50mM Sodium Borate |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human Phospholipase C delta 1 fragment. |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?