missing translation for 'onlineSavingsMsg'
Learn More

Phospholipase B1 Antibody (OTI1B3), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18430187 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430187 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430187 Supplier Novus Biologicals Supplier No. NBP273385AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Phospholipase B1 Monoclonal antibody specifically detects Phospholipase B1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospholipase B1
Applications Western Blot
Classification Monoclonal
Clone OTI1B3
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias FLJ30866, hPLB, phospholipase B1, PLBmembrane-associated
Host Species Mouse
Immunogen Full length recombinant protein of human Phospholipase B1 (NP_694566) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 151056
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.