missing translation for 'onlineSavingsMsg'
Learn More

Phospholipase A2 X Antibody (5G11), Novus Biologicals™

Product Code. 18386198 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386198 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386198 Supplier Novus Biologicals Supplier No. H00008399M01100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Phospholipase A2 X Monoclonal antibody specifically detects Phospholipase A2 X in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospholipase A2 X
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5G11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_003552.1
Gene Alias EC 3.1.1.4, group 10 secretory phospholipase A2, Group X secretory phospholipase A2, GX sPLA2, GXPLA2, GXSPLA2, MGC119918, MGC119919, MGC133367, Phosphatidylcholine 2-acylhydrolase 10, phospholipase A2, group X, SPLA2, sPLA2-X
Host Species Mouse
Immunogen PLA2G10 (NP_003552.1, 64 a.a. ~ 165 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KYGCFCGLGGHGQPRDAIDWCCHGHDCCYTRAEEAGCSPKTERYSWQCVNQSVLCGPAENKCQELLCKCDQEIANCLAQTEYNLKYLFYPQFLCEPDSPKCD
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 8399
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.