missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholipase A2 IID Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93007-0.02ml
This item is not returnable.
View return policy
Description
Phospholipase A2 IID Polyclonal antibody specifically detects Phospholipase A2 IID in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Phospholipase A2 IID | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:10-1:100, Immunohistochemistry-Paraffin | |
| EC 3.1.1.4, GIID sPLA2, group IID secretory phospholipase A2, Phosphatidylcholine 2-acylhydrolase 2D, phospholipase A2, group IID, PLA2IID, secretory phospholipase A2s, Secretory-type PLA, stroma-associated homolog, sPLA2S, SPLASHsPLA2-IID | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 21-145 of human PLA2G2D (NP_036532.1). GILNLNKMVKQVTGKMPILSYWPYGCHCGLGGRGQPKDATDWCCQTHDCCYDHLKTQGCSIYKDYYRYNFSQGNIHCSDKGSWCEQQLCACDKEVAFCLKRNLDTYQKRLRFYWRPHCRGQTPGC | |
| 0.02 mL | |
| Lipid and Metabolism | |
| 26279 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction