missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholemman/FXYD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13721-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the FXYD family of small membrane proteins that share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD and containing 7 invariant and 6 highly conserved amino acids. The protein encoded by this gene is a plasma membrane substrate for several kinases, including protein kinase A, protein kinase C, NIMA kinase, and myotonic dystrophy kinase. It is thought to form an ion channel or regulate ion channel activity and act as an accessory protein of Na, K-ATPase. Alternatively spliced transcript variants have been described.Specifications
| Phospholemman/FXYD1 | |
| Polyclonal | |
| Unconjugated | |
| FXYD1 | |
| FXYD1, Phospholemman, PLM | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5348, 56188, 58971 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00168, O08589, Q9Z239 | |
| Fxyd1 | |
| Phospholemman/FXYD1 Fusion Protein Ag4669 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction