missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholamban Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £376.00
Specifications
| Antigen | Phospholamban |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18393503
|
Bio-Techne
NBP3-16904-20UL |
20 μg |
£149.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18356682
|
Bio-Techne
NBP3-16904-100UL |
100 μg |
£376.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospholamban Polyclonal antibody specifically detects Phospholamban in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Phospholamban | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific, Signal Transduction | |
| PBS, 50% glycerol, pH7.3 | |
| 5350 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CMD1PPLBcardiac phospholamban, phospholamban | |
| A synthetic phosphorylated peptide around S16 & T17 of human Phospholamban (NP_002658.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title