missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospholamban Polyclonal Antibody
GREENER_CHOICE

Product Code. 16382205
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16382205 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16382205 Supplier Invitrogen™ Supplier No. PA585268

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (98%), Rat (98%), Dog (96%), Pig (96%), Rabbit (98%), Chicken (82%), Rhesus Monkey (98%), Bovine (94%). Positive Control: mouse heart. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Phospholamban (PLB) is a 52 amino acid phosphoprotein which regulates the calcium pump of cardiac sarcoplasmic reticulum (SR). PLB is an oligomer of five identical subunits each having a cytoplasmic and transmembrane domain. The cytoplasmic domain (residues 1 to 25) contains the phosphorylation sites and is highly basic and readily cleaved by proteases whereas the transmembrane domain (residues 25 to 52) is mostly hydrophobic, protease resistant and stabilizes the pentamer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospholamban
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene PLN
Gene Accession No. P26678, P61014, P61016
Gene Alias cardiac CMH18; Cardiac phospholamban; CMD1P; CMH18; phospholamban; Plb; Plm; PLN; unnamed protein product
Gene Symbols PLN
Host Species Rabbit
Immunogen Full length human Phospholamban Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18821, 5350, 64672
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.