missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholamban Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97463PCP
This item is not returnable.
View return policy
Description
Phospholamban Polyclonal antibody specifically detects Phospholamban in Human, Cynomolgus Monkey samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Phospholamban | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| CMD1PPLBcardiac phospholamban, phospholamban | |
| Produced in rabbits immunized with E. coli-derived Human Phospholamban fragment. | |
| 0.1 mL | |
| Phospho Specific, Signal Transduction | |
| 5350 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human, Cynomolgus Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction