missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoglucomutase 5 Mouse anti-Human, Mouse, Rat, Bovine, Rabbit, Clone: 14F8/F8, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07812-100ug
This item is not returnable.
View return policy
Description
Phosphoglucomutase 5 Monoclonal specifically detects Phosphoglucomutase 5 in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Phosphoglucomutase 5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin | |
| aciculin, EC 5.4.2.2, PGM-RP, PGMRPphosphoglucomutase-like protein 5, phosphoglucomutase 5, Phosphoglucomutase-related protein | |
| Cytoskeletal fraction of the human myometrium tissue extract | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Bovine, Rabbit | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 14F8/F8 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5239 | |
| Store at 4°C. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction