missing translation for 'onlineSavingsMsg'
Learn More

Phosphoglucomutase 5 Antibody (PGM5/7005R), FITC, Novus Biologicals™

Product Code. 18322945 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322945 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322945 Supplier Novus Biologicals Supplier No. NBP314134F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Phosphoglucomutase 5 Monoclonal antibody specifically detects Phosphoglucomutase 5 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphoglucomutase 5
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PGM5/7005R
Conjugate FITC
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias aciculin, EC 5.4.2.2, PGM-RP, PGMRPphosphoglucomutase-like protein 5, phosphoglucomutase 5, Phosphoglucomutase-related protein
Host Species Rabbit
Immunogen Cytoskeletal fraction of the human myometrium tissue extract (Uniprot: Q15124)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5239
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.