missing translation for 'onlineSavingsMsg'
Learn More

Phosphoglucomutase 5 Antibody (PGM5/3552) - Azide and BSA Free, Novus Biologicals™

Product Code. 18325936 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325936 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325936 Supplier Novus Biologicals Supplier No. NBP314132

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Phosphoglucomutase 5 Monoclonal antibody specifically detects Phosphoglucomutase 5 in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphoglucomutase 5
Applications Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone PGM5/3552
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 to 2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array
Formulation 10mM PBS without Preservative
Gene Alias aciculin, EC 5.4.2.2, PGM-RP, PGMRPphosphoglucomutase-like protein 5, phosphoglucomutase 5, Phosphoglucomutase-related protein
Host Species Mouse
Immunogen Recombinant full-length human Phosphoglucomutase 5 protein (Uniprot: Q15124)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5239
Target Species Human, Mouse, Rat, Bovine, Rabbit
Content And Storage Store at -20 to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.