missing translation for 'onlineSavingsMsg'
Learn More

Phosphoenolpyruvic Acid, Monopotassium Salt, CDN

Product Code. 30327473
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30327473 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30327473 Supplier CDN Supplier No. CDNX34301G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phospho(enol)pyruvic Acid Monopotassium Salt
CAS 4265-07-0
Molecular Formula C3 H4 O6 P . K
Quantity 1 g
Synonym 2-Propenoic acid, 2-(phosphonooxy)-, potassium salt (1:1), 2-Propenoic acid, 2-(phosphonooxy)-, monopotassium salt (9 CI), Acrylic acid, 2-hydroxy-, dihydrogen phosphate, monopotassium salt (8 CI), Monopotassium phosphoenolpyruvate, Phosphoenolpyruvic acid potassium salt, 2-(Phosphonooxy)-2-propenoic acid monopotassium salt, Phosphoenolpyruvic acid monopotassium salt, Phospho(enol)pyruvic acid monopotassium salt
InChI Formula InChI=1 S/C3H5O6P.K/c1-2(3(4)5)9-10(6,7)8;/h1H2,(H,4,5)(H2,6,7,8);/q;+1/p-1
SMILES [K+].OC(=O)C(=C)OP(=O)(O)[O-]
IUPAC Name potassium;1-carboxyethenyl hydrogen phosphate
Molecular Weight (g/mol) 206.132
Formula Weight 205.938 g/mol
Percent Purity min 98% chemical purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.