missing translation for 'onlineSavingsMsg'
Learn More

Phosphoenolpyruvic acid mono(cyclohexylammonium) salt, 98%

Product Code. 11409543
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409543 25 g 25g
11489533 1 g 1g
11499533 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409543 Supplier Thermo Scientific Alfa Aesar Supplier No. A12385.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10526-80-4 | C9H18NO6P | 267.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10526-80-4
Molecular Formula C9H18NO6P
Molecular Weight (g/mol) 267.22
MDL Number MFCD00036375
InChI Key VHFCNZDHPABZJO-UHFFFAOYSA-N
Synonym cyclohexanamine 2-phosphonooxy acrylate, phosphoenolpyruvic acid cyclohexylammonium salt, 2-propenoic acid, 2-phosphonooxy-, compd. with cyclohexanamine 1:1, cyclohexylamine; phosphoenolpyruvic acid, phosphoenolpyruvic acid cyclohexylamine salt, phosphoenolpyruvic acid, cyclohexylammonium salt, cyclohexanamine; 2-phosphonooxyprop-2-enoic acid, phospho enol pyruvic acid cyclohexylammonium salt, phosphoenolpyruvic acid monocyclohexylammonium salt
PubChem CID 82702
IUPAC Name cyclohexanamine;2-phosphonooxyprop-2-enoic acid
SMILES NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O

Specifications

Melting Point ∼140°C (decomposition)
Quantity 25 g
Formula Weight 267.22
Percent Purity 98%
Chemical Name or Material Phosphoenolpyruvic acid mono (cyclohexylammonium) salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.