missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoenolpyruvate Carboxylase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44235
This item is not returnable.
View return policy
Description
Specifically detects Phospho Enol Pyruvate Carboxylase in C. elegans, Plant samples, and it is validated for ELISA, Immunohistochemistry, Western Blotting
Phosphoenolpyruvate Carboxylase Polyclonal specifically detects Phosphoenolpyruvate Carboxylase in Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Phosphoenolpyruvate Carboxylase | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| PEPC, PEPC 1, PEPC1, PEPCase, PEPCase 1, Phosphoenolpyruvate carboxylase 1, PPC1, PPEPC4 | |
| Rabbit | |
| 109.36 kDa | |
| 0.1 mg | |
| Primary | |
| Reconstitute with 100 ul deionized water (or equivalent) | |
| Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1:1000-1:4000, ELISA 1:10000-1:40000, Immunohistochemistry 1:10-1:500 | |
| B8XPZ2 | |
| LOC542372 | |
| Phosphoenolpyruvate Carboxylase Antibody was produced by repeated immunizations with maize leaves Phospho-enol-pyruvate Carboxylase. (Uniprot: B8XPZ2) | |
| Multi-step | |
| RUO | |
| 542372 | |
| Plant | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction