missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Phosphodiesterase I (C. adamanteus)
Click to view available options
:
100 Units
Description
#REF!
Specifications
Specifications
| For Use With (Application) | Enzyme Activity |
| Molecular Weight (g/mol) | 115kDa |
| Name | Phosphodiesterase I (C. adamanteus) |
| Preparation Method | Native protein purified from Crotalus adamanteus Venom according to the method of Williams, Sung and Laskowski, J. Biol. Chem., 236, 1130 (1961). Further treated to inactivate contaminating 5'-nucleotidase activity according to Sulkowski and Laskowski: Biochim. Biophys. Acta, 240, 443 (1961). |
| Quantity | 100 Units |
| Storage Requirements | Store at -20°C. |
| Regulatory Status | RUO |
| Common Name | PHOSPHODIESTERASE |
| Assays | Phosphodiesterase I Assay; Structural and sequence studies of nucleic acids. |
| Biological Activity | ≥20 units per mg dry weight. One Unit hydrolyzes one micromole of p-nitrophenyl thymidine-5-phosphate per minute at 25°C, pH 8.9. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction