missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphodiesterase 4D Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-2462
This item is not returnable.
View return policy
Description
Phosphodiesterase 4D Polyclonal specifically detects Phosphodiesterase 4D in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| Phosphodiesterase 4D | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E3), cAMP-specific 3'-5'-cyclic phosphodiesterase 4D, cAMP-specific phosphodiesterase PDE4D6, DKFZp686M11213, Drosophila), dunce-like phosphodiesterase E3, EC 3.1.4, FLJ97311, HSPDE4D, phosphodiesterase 4D, cAMP-specific, phosphodiesterase 4D, cAMP-specific (phosphodiesterase E3 dunce homolog | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antiobody is expected to recognize all reported isoforms. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/ml, ELISA 1:200, Immunohistochemistry-Paraffin 5 ug/ml, Peptide ELISA Detection limit 1:128000 | |
| NP_006194.2 | |
| PDE4D | |
| Peptide with sequence QPEACVIDDRSPDT corresponding to C-Terminus according to NP_001184151.1, NP_001184150.1, NP_006194.2, NP_001098101.1, NP_001184152.1, NP_001159371.1. | |
| 0.1 mg | |
| Lipid and Metabolism, Signal Transduction | |
| 5144 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction