missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphodiesterase 4A/PDE4A, p Ser686, p Ser688 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £443.00
Specifications
| Antigen | Phosphodiesterase 4A/PDE4A |
|---|---|
| Dilution | Western Blot 1:500, ELISA 1:10000, Immunoprecipitation 1:150 |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18201195
|
Novus Biologicals
NB300-635-0.025MG |
0.025 mg |
£292.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18274374
|
Novus Biologicals
NB300-635-0.05MG |
0.05 mg |
£443.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phosphodiesterase 4A/PDE4A Polyclonal specifically detects Phosphodiesterase 4A/PDE4A in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
| Phosphodiesterase 4A/PDE4A | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Human, Mouse, Rat, Rabbit | |
| P27815 | |
| 5141 | |
| Synthetic peptide corresponding to PKA phosphorylation site on PDE4A long form enzymes. The peptide was synthesized with phosphorylated serine. | |
| Primary | |
| This detects all known long-from variants of PDE4A, including PDE4A4, PDE4A5, PDE4A8, PDE4Ax when phosphorylated at the PKA phophorylation site. It will not cross-react with PDE4A phosphorylated at the ERK phosphorylation site. It does not cross-react with PDE4B, PDE4D or any other member of the PDE family. |
| Western Blot 1:500, ELISA 1:10000, Immunoprecipitation 1:150 | |
| Polyclonal | |
| Rabbit | |
| Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide | |
| cAMP-specific 3′-5′-cyclic phosphodiesterase 4A, DPDE2cAMP-specific phosphodiesterase, EC 3.1.4, EC 3.1.4.17, PDE4, PDE46, phosphodiesterase 4A, cAMP-specific, phosphodiesterase 4A, cAMP-specific (dunce, phosphodiesterase 4A, cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E2), phosphodiesterase E2 dunce homolog, Drosophila, phosphodiesterase isozyme 4 | |
| PDE4A | |
| IgG | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title