missing translation for 'onlineSavingsMsg'
Learn More

Phosphodiesterase 4A/PDE4A Antibody (OTI1C8), FITC, Novus Biologicals™

Product Code. 18120917 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18120917 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18120917 Supplier Novus Biologicals Supplier No. NBP273376F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Phosphodiesterase 4A/PDE4A Monoclonal specifically detects Phosphodiesterase 4A/PDE4A in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphodiesterase 4A/PDE4A
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1C8
Conjugate FITC
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias cAMP-specific 3′-5′-cyclic phosphodiesterase 4A, DPDE2cAMP-specific phosphodiesterase, EC 3.1.4, EC 3.1.4.17, PDE4, PDE46, phosphodiesterase 4A, cAMP-specific, phosphodiesterase 4A, cAMP-specific (dunce, phosphodiesterase 4A, cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E2), phosphodiesterase E2 dunce homolog, Drosophila, phosphodiesterase isozyme 4
Gene Symbols PDE4A
Host Species Mouse
Immunogen Full length recombinant protein of human Phosphodiesterase 4A/PDE4A (NP_006193) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5141.0
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.