missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHOSPHO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584679
This item is not returnable.
View return policy
Description
Immunogen sequence: RDLSAIYEAI PLSPGMSDLL QFVAKQGACF EVILISDANT FGVESSLRAA GHHSLFRRIL SNPSGPDARG LLALRPF Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 90%.
Phosphatase that has a high activity toward phosphoethanolamine (PEA) and phosphocholine (PCho). Involved in the generation of inorganic phosphate for bone mineralization. [UniProt].
Specifications
| PHOSPHO1 | |
| Polyclonal | |
| Unconjugated | |
| PHOSPHO1 | |
| D11Moh36; phosphatase, orphan 1; PHOSPHO1; Phosphoethanolamine/phosphocholine phosphatase; Phospo1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 162466 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TCT1 | |
| PHOSPHO1 | |
| Recombinant protein corresponding to Human PHOSPHO1. Recombinant protein control fragment (Product #RP-108505). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction