missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-ZIPK (Thr265) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540234
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
DAPK3 is a serine/threonine kinase which is functionally linked to cell motility, smooth muscle contractility and cell death processes. DAPK3 is encoded by ZIPK and is a component of desmosomes and of the epidermal cornified envelope in keratinocytes. The N-terminal domain of this protein interacts with the plasma membrane and its C-terminus interacts with intermediate filaments. Through its rod domain, this protein forms complexes with envoplakin. This protein may serve as a link between the cornified envelope and desmosomes as well as intermediate filaments. AKT1/PKB, a protein kinase mediating a variety of cell growth and survival signaling processes, is reported to interact with this protein, suggesting a possible role for this protein as a localization signal in AKT1-mediated signaling.
Specifications
| Phospho-ZIPK (Thr265) | |
| Polyclonal | |
| Unconjugated | |
| DAPK3 | |
| DAP kinase 3; DAP3K; Dapk3; Dapkl; DAP-like kinase; death associated protein kinase 3; death-associated kinase 3; Death-associated like kinase; death-associated protein kinase 3; DLK; MYPT1 kinase; ZIP; ZIP kinase; ZIP kinase isoform; Zipk; ZIP-kinase; zipper-interacting protein kinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13144, 1613, 64391 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43293, O54784, O88764 | |
| DAPK3 | |
| Synthesized peptide derived from human DAPK3 around the phosphorylation site of T265. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction