missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-ZAP70/Syk (Tyr319, Tyr352) Monoclonal Antibody (n3kobu5), APC, eBioscience™

Product Code. 15508216
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15508216 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15508216 Supplier Invitrogen™ Supplier No. 17900642

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Description: This n3kobu5 monoclonal antibody recognizes human and mouse zeta chain-associated protein of 70 kD (also known as ZAP-70) and spleen tyrosine kinase (also known as SYK) when phosphorylated on Y319 and Y352, respectively. ZAP-70 and SYK are members of the SYK protein tyrosine kinase (PTK) family that are phosphorylated and activated by Src family PTKs. ZAP-70/SYK Y319/Y352 are located in the so-called interdomain of ZAP-70/SYK (between the N-terminal dual SH2 domains and the C-terminal kinase domain). Phosphorylation of ZAP-70 Y319 by Lck is necessary for T cell receptor (TCR)-dependent association of ZAP-70 with Lck and phospholipase C gamma and subsequent activation of calcium-dependent and Ras signaling cascades. SYK Y352 phosphorylation by Fyn/Lyn is critical for propagation of B cell receptor (BCR) signaling and for B cell development. Specificity of this n3kobu5 clone was determined by ELISA, flow cytometry, and western blotting. Applications Reported:This n3kobu5 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This n3kobu5 antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically b...

ZAP-70 is a cytosolic protein tyrosine kinase that is a member of the Syk family of proteins. ZAP-70 is expressed exclusively in T-cells and natural killer cells and is required for T-cell receptor activation. Upon T cell antigen receptor (TCR) engagement, ZAP70 is phosphorylated on tyrosines 292, 315 and 319 in the interdomain B, located between the SH2 and kinase domains. Phosphorylation of both tyrosines 315 (a Vav binding site) and 319 (a Lck binding site) enhances ZAP70 function in mediating lymphocyte signaling, while tyrosine 292 terminates the transient activation of ZAP70 and attentuates lymphocyte signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-ZAP70/Syk (Tyr319, Tyr352)
Applications Flow Cytometry
Classification Monoclonal
Clone n3kobu5
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene ZAP70
Gene Accession No. P43403, P43404
Gene Alias 70 kDa zeta-associated protein; 70 kDa zeta-chain associated protein; ADMIO2; EC 2.7.10.2; FLJ17670; FLJ17679; I79_004087; IMD48; kinase ZAP70; mrtle; mur; Selective T cell defect; Srk; STCD; STD; syk-related protein tyrosine kinase; syk-related tyrosine kinase; Tyrosine-protein kinase ZAP-70; Tyrosine-protein kinase ZAP-70;TZK; TZK; ZA70; Zap70; ZAP-70; Zeta chain associated protein kinase 70kDa; zeta chain of T cell receptor associated protein kinase; zeta chain of T cell receptor associated protein kinase 70; zeta chain of T cell receptor associated protein kinase 70kDa; zeta-chain (TCR) associated protein kinase; zeta-chain (TCR) associated protein kinase 70; zeta-chain (TCR) associated protein kinase 70kDa; zeta-chain associated protein kinase 70kDa; zeta-chain associated protein kinase, 70kD
Gene Symbols ZAP70
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22637, 7535
Target Species Human, Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.