missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-YAP1 (Ser127) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114675
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of YAP only when phosphorylated at Serine 127.
YAP (Yes-Associated Protein) is a founding member of the WW domain family of proteins, transcriptional co-activator and oncogene. Its gene is transcriptionally activated in squamous cell carcinoma cells. YAP can act both as a coactivator and corepressor and is the critical downstream regulatory target in the Hippo signaling pathway. The Hippo signaling pathway plays a pivotal role in organ size control and tumour suppression by restricting proliferation and promoting apoptosis. Also plays key roles in tissue tension, 3D tissue shape, cell proliferation, cell death, and cell migration. Mutations in the IRF5 gene results in coloboma.
Specifications
| Phospho-YAP1 (Ser127) | |
| Polyclonal | |
| Unconjugated | |
| YAP1 | |
| 65 kDa Yes-associated protein; AI325207; cb194; CH211-181P1.5-001; COB1; Protein yorkie homolog; sb:cb194; si:ch211-181p1.5; si:dkey-3b8.3; transcriptional coactivator YAP1; wu:fc18c04; yap; Yap1; YAP2; Yap65; Yes associated protein 1; Yes1 associated transcriptional regulator; yes-associated protein 1; yes-associated protein 2; yes-associated protein YAP65 homolog; yes-associated protein, 65 kDa; Yki; Yorkie; yorkie homolog; zgc:158380; zYAP | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 10413, 22601, 363014 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P46937, P46938, Q2EJA0 | |
| YAP1 | |
| A synthesized peptide derived from human YAP1(Accession P46937), corresponding to amino acid residues around phosphorylated Ser127. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction