missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Vinculin (Tyr692) Polyclonal Antibody

Phospho-Vinculin (Tyr692) Polyclonal Antibody
Brand: Invitrogen PA5105518
This item is not returnable.
View return policy
Description
Phospho-Vinculin (Tyr692) Polyclonal Antibody for Western Blot
Specifications
| Phospho-Vinculin (Tyr692) | |
| Polyclonal | |
| Unconjugated | |
| VCL | |
| 9430097D22; AA571387; AI462105; AW545629; CMD1W; CMH15; epididymis luminal protein 114; HEL114; metavinculin; meta-vinculin; metavinculin {ECO:0000250; MV; MVCL; RP11-178G16.3; UniProtKB:P18206}; unnamed protein product; Vcl; vcl {ECO:0000250; VIN; vinc; vinc 1; VINC1; Vinculin; vinculin {ECO:0000250 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 22330, 305679, 7414 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P18206, P85972, Q64727 | |
| VCL | |
| A synthesized peptide derived from human VCL(Accession P18206), corresponding to amino acid residues around phosphorylated Tyr692. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction