missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Vimentin (Ser39) Recombinant Rabbit Monoclonal Antibody (3H9L2)
Phospho-Vimentin (Ser39) Recombinant Rabbit Monoclonal Antibody (3H9L2)
Brand: Invitrogen 702179
This item is not returnable.
View return policy
Description
Phospho-Vimentin (Ser39) Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Specifications
| Phospho-Vimentin (Ser39) | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P08670 | |
| Vim | |
| Peptide corresponding to Human VIM (aa 36-45). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 3H9L2 | |
| Unconjugated | |
| Vim | |
| cb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; HEL113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminus | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7431 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction