missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-VGAT (Ser462) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105326
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of SLC32A1 only when phosphorylated at Ser462.
The Vesicular GABA Amino Acid Transporter (VGAT) is responsible for transport of the inhibitory neurotransmitter into synaptic vesicles (McIntire et al., 1997). The VGAT protein (also known as the Vesicular Inhibitory Amino Aid Transporter or VIAAT) is expressed in synaptic vesicles of both glycine and GABAergic synapses throughout the CNS (Chaudhry et al., 1998). Expression of the VGAT protein changes during development and also in response to patterns of neuronal activity (De et al., 2005).
Specifications
| Phospho-VGAT (Ser462) | |
| Polyclonal | |
| Unconjugated | |
| SLC32A1 | |
| GABA and glycine transporter; GVAT; hVIAAT; mVGAT; mVIAAT; R75019; rGVAT; SLC32A1; solute carrier family 32 (GABA vesicular transporter), member 1; solute carrier family 32 member 1; vasicular inhibitory amino acid transporter 10D; vesicular GABA transporter; Vesicular inhibitory amino acid transporter; Vgat; Viaat | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 140679, 22348, 83612 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35458, O35633, Q9H598 | |
| SLC32A1 | |
| A synthesized peptide derived from human SLC32A1(Accession Q9H598), corresponding to amino acid residues around phosphorylated Ser462. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction