missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-VASP (Ser239) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117182
This item is not returnable.
View return policy
Description
Positive Samples: 293T Immunogen sequence: KVSKQ.
Vasodilator-stimulated phosphoprotein (VASP) is a member of the Ena-VASP protein family. Ena-VASP family members contain an EHV1 N-terminal domain that binds proteins containing E/DFPPPPXD/E motifs and targets Ena-VASP proteins to focal adhesions. In the mid-region of the protein, family members have a proline-rich domain that binds SH3 and WW domain-containing proteins. Their C-terminal EVH2 domain mediates tetramerization and binds both G and F actin. VASP is associated with filamentous actin formation and likely plays a widespread role in cell adhesion and motility. VASP may also be involved in the intracellular signaling pathways that regulate integrin-extracellular matrix interactions. VASP is regulated by the cyclic nucleotide-dependent kinases PKA and PKG.
Specifications
| Phospho-VASP (Ser239) | |
| Polyclonal | |
| Unconjugated | |
| VASP | |
| Enah/Vasp-like; I79_012660; Unknown (protein for MGC:127789); vasodilator stimulated phosphoprotein; vasodilator-stimulated phosphoprotein; vasodilator-stimulated phosphoprotein (VASP); VASP | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22323, 361517, 7408 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.87 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P50552, P70460 | |
| VASP | |
| A synthetic phosphorylated peptide around S239 of human VASP (NP_0033611). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction