missing translation for 'onlineSavingsMsg'
Learn More

Phospho-VASP (Ser238) Polyclonal Antibody, Invitrogen™

Product Code. 15515081
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15515081 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15515081 Supplier Invitrogen Supplier No. PA538152

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VASP (vasodilator-stimulated phosphoprotein), is involved in the maintece of cytoarchitecture by interacting with actin-like filaments. VASP shares a limited degree of homology with the amino terminus of WASP, which is frequently mutated in WAS patients. An established substrate of cAMP and cGMP dependent kinases, VASP is phosphorylated on a regulatory serine residue 157 and localizes to focal adhesions, microfilaments and highly active regions of the plasma membrane. VASP is a protein of between 46 and 50 kDa that is highly expressed in human platelets and, like WASP, may play a role in cytoskeletal organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-VASP (Ser238)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene VASP
Gene Accession No. P50552, P70460
Gene Alias VASP, Vasodilator-stimulated phosphoprotein
Gene Symbols VASP
Host Species Rabbit
Immunogen A synthetic phosphopeptide derived from human VASP around the phosphorylation site of Ser238 (K-V-SP-K-Q)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22323, 361517, 7408
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.