missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-VANGL2 (Ser11, Ser14) Recombinant Rabbit Monoclonal Antibody (4D3W2)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542956
This item is not returnable.
View return policy
Description
The target is usually found in the following locations: Cell membrane, Multi-pass membrane protein.
Van Gogh is an alternate name for the Drosophila strabismus protein. Van Gogh like-2 (VANGL2) is a 521 amino acidprotein in vertebrates similar to strabismus 1 of Drosophila and a component of the frizzled-disheveled tissue polarity pathway. It interacts through it's C-terminal region with the N-terminal half of DVL1, DVL2 and DVL3 and is a potent tumor suppressor. It ispredominantly involved in the control of early morphogenesis and patterning of both axial midline structures and the development of neural plate. It plays a crucial role in the regulation of planar cell polarity, particularly in the orientation of stereociliary bundles in the cochlea. VANGL2 is required for polarization and movement of myocardializing cells and seems to act via RHOA signaling to regulate this process. It is expressed in a wide range of tissues.
Specifications
| Phospho-VANGL2 (Ser11, Ser14) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q91ZD4, Q9ULK5 | |
| Vangl2 | |
| A phospho specific peptide corresponding to residues surrounding S11/S14 of human VANGL2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 4D3W2 | |
| Unconjugated | |
| Vangl2 | |
| C530001F03Rik; KIAA1215; loop tail associated protein; loop-tail; Loop-tail protein 1; loop-tail protein 1 homolog; Loop-tail-associated protein; Lp; LPP1; Ltap; MGC119403; MGC119404; RGD:1309442}; ska>m17Jus>; ska17; STB1; stbm; STBM1; strabismus; Strabismus 1; van Gogh-like protein 2; vang; vang (van gogh)-like 2; Vang1l2; VANGL planar cell polarity protein 2; Vangl2; vangl2 {ECO:0000312; vang-like 2 (van gogh, Drosophila); vang-like protein 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57216, 93840 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction