missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-VAMP4 (Ser30) Polyclonal Antibody

Product Code. 15752606
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15752606 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15752606 Supplier Invitrogen™ Supplier No. PA564540

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Phospho-VAMP4 (Ser30) Polyclonal Antibody detects endogenous levels of VAMP4 only when phosphorylated at Ser30.

The vesicle associated membrane proteins (VAMP) or synaptobrevins are calcium binding proteins specific to eukaryotes. VAMPs, along with synaptosomal associated protein of 25 kDa (SNAP-25) and syntaxin, form the core complex of soluble NSF attachment protein receptor (SNARE) proteins that interact with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and alpha-SNAP. These membrane associated proteins play a key role in the regulation of vesicle membrane fusion with the plasma membrane. The Clostridium tetani neurotoxin is a metalloprotease with specificity for VAMP. In Alzheimer's disease, VAMP levels of all isoforms appear to be significantly lowered.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-VAMP4 (Ser30)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene VAMP4
Gene Accession No. O70480, O75379
Gene Alias D1Ertd147e; vamp; VAMP24; VAMP4; VAMP-4; vesicle associated membrane protein 4; vesicle-associated membrane protein 4
Gene Symbols VAMP4
Host Species Rabbit
Immunogen A synthesized peptide derived from human VAMP4 (Phospho-Ser30).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 364033, 53330, 8674
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.