missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-ULK1 (Thr1046) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105423
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of ULK1 only when phosphorylated at Thr1046.
ULK1 is a serine/threonine protein kinase that plays critical role during initial stages of autophagy which is a vital response to nutrient starvation. The conserved C-terminal domain (CTD) of ULK1 controls the regulatory function and localization of the protein. Knockdown of ULK1 inhibits the autophagic response as well as inhibiting rapamycin-induced autophagy consistent with a role downstream of mTOR. ULK1 forms a complex with FIP200 and ATG13 and this complex is essential for starvation-induced autophagy. Both FIP200 and ATG13 are critical for correct localization of ULK1 to the pre-autophagosome and stability of ULK1 protein. ULK1 is phosphorylated by the mTOR pathway in a nutrient starvation-regulated manner.
Specifications
| Phospho-ULK1 (Thr1046) | |
| Polyclonal | |
| Unconjugated | |
| ULK1 | |
| APG1; ATG 1; ATG1; ATG1 autophagy related 1 homolog; ATG1A; AU041434; Autophagy-related protein 1 homolog; C. elegans; EC:2.7.11.1; FLJ38455; hATG1; KIAA0722; mKIAA0722; Serine/threonine-protein kinase ULK1; serine/threonine-protein kinase Unc51.1; ULK 1; Ulk1; UNC 51; UNC51; unc-51 like autophagy activating kinase 1; unc-51 like kinase 1; Unc51.1; unc-51-like kinase 1 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 22241, 360827, 8408 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O70405, O75385 | |
| ULK1 | |
| A synthesized peptide derived from human ULK1(Accession O75385), corresponding to amino acid residues around phosphorylated Thr1046. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction