missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Tyrosine Antibody (PY2870R) - Azide and BSA Free, Novus Biologicals™

Product Code. 18393962 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393962 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393962 Supplier Novus Biologicals Supplier No. NBP308423

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Phospho-Tyrosine Monoclonal specifically detects Phospho-Tyrosine in All Species samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Tyrosine
Applications ELISA
Classification Monoclonal
Clone PY2870R
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 10mM PBS
Gene Alias 2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, phosphotyrosine, pTyrosine, pY20, Tyrosine
Host Species Rabbit
Immunogen Recombinant full-length human Phospho-Tyrosine protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.